Learn More
Medchemexpress LLC Cefetamet | 65052-63-3 | 99.4% | 397.43 | C14H15N5O5S2 | 10 MG
SDP
Catalog No. 5000298316
Encompass_Preferred
Cefetamet is a cephalosporin antibiotic research compound that acts as the active metabolite of cefetamet pivoxil. It binds bacterial penicillin-binding proteins and exhibits activity against a range of Gram-negative and Gram-positive bacteria. It is supplied as a solid in small-mass packs and as DMSO solutions for assay use.
  • Cephalosporin antibiotic with defined antibacterial activity.
  • Active metabolite used in studies of penicillin-binding proteins.
  • High purity suitable for biochemical and microbiological research.
  • Offered in small solid packs and 10 mM DMSO solutions for assays.
  • Well characterized by CAS number 65052-63-3 and molecular weight 397.43.
Catalog No. 50-002-98316 Supplier Medchemexpress LLC Supplier No. HYA011110MG
Cefetamet is a cephalosporin antibiotic research compound that acts as the active metabolite of cefetamet pivoxil. It binds bacterial penicillin-binding proteins and exhibits activity against a range of Gram-negative and Gram-positive bacteria. It is supplied as a solid in small-mass packs and as DMSO solutions for assay use.
  • Cephalosporin antibiotic with defined antibacterial activity.
  • Active metabolite used in studies of penicillin-binding proteins.
  • High purity suitable for biochemical and microbiological research.
  • Offered in small solid packs and 10 mM DMSO solutions for assays.
  • Well characterized by CAS number 65052-63-3 and molecular weight 397.43.