Learn More
Selleck Chemical LLC Cefetamet pivoxil hydrochloride S4865-25mg
SDP
Supplier:  Selleck Chemical LLC S486525MG
Encompass_Preferred
Cefetamet pivoxil hydrochloride is the hydrochloride salt form of cefetamet pivoxil which is an oral third-generation cephalosporin antibiotic
Catalog No. 50-311-2661