Learn More
Medchemexpress LLC Cefetamet (Ro 15-8074) | 65052-63-3 | 99.1% | C14H15N5O5S2 | 50 MG
SDP
Catalog No. 5000432466
Encompass_Preferred
Cefetamet (Ro 15-8074) is a cephalosporin antibiotic and the active metabolite of Cefetamet pivoxil. It functions by binding to bacterial penicillin-binding protein (PBP), with an IC50 for PBP3 in *Escherichia coli* W3110 of 2.5 μg/mL.
  • Demonstrates significant activity against various Gram-negative bacteria (including Enterobacteriaceae, Neisseria species, and Haemophilus influenzae) and Gram-positive bacteria such as Streptococcus.
  • Exhibits high in vitro activity against most members of Enterobacteriaceae, Haemophilus influenzae, Neisseria spp., and Streptococcus pneumoniae (MIC90 ≤ 2 μg/mL).
  • Kills and lyses Treponema pallidum.
  • Can be utilized in research for respiratory tract, urinary tract, ear, nose, and throat infections, and syphilis.
  • In vivo studies show it can cure experimental syphilis in rabbits (1-30 mg/kg; i.m.; once).
Catalog No. 50-004-32466 Supplier Medchemexpress LLC Supplier No. HYA011150MG
Cefetamet (Ro 15-8074) is a cephalosporin antibiotic and the active metabolite of Cefetamet pivoxil. It functions by binding to bacterial penicillin-binding protein (PBP), with an IC50 for PBP3 in *Escherichia coli* W3110 of 2.5 μg/mL.
  • Demonstrates significant activity against various Gram-negative bacteria (including Enterobacteriaceae, Neisseria species, and Haemophilus influenzae) and Gram-positive bacteria such as Streptococcus.
  • Exhibits high in vitro activity against most members of Enterobacteriaceae, Haemophilus influenzae, Neisseria spp., and Streptococcus pneumoniae (MIC90 ≤ 2 μg/mL).
  • Kills and lyses Treponema pallidum.
  • Can be utilized in research for respiratory tract, urinary tract, ear, nose, and throat infections, and syphilis.
  • In vivo studies show it can cure experimental syphilis in rabbits (1-30 mg/kg; i.m.; once).