Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cefiderocol Antimicrobial Susceptibility Discs

Catalog No. CT1964B Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
30 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
CT1964B 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CT1964B Supplier Thermo Scientific™ Supplier No. CT1964B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform manual susceptibility testing of multi-drug and extensively drug resistant bacteria with AST discs which meet EUCAST and CLSI breakpoints

Manually determine the antibiotic susceptibility of microorganisms using disc diffusion methods with consistently reliable, industry-leading quality1 Thermo Scientific™ Oxoid™ Cefiderocol Antimicrobial Susceptibility Discs.

The drug cefiderocol has been designed to address infections caused by aerobic Gram-negative organisms that are multi-drug (MDR) or extensively drug (XDR) resistant i.e. for adults who have limited treatment options. Cefiderocol is aimed at the treatment of serious infections, such as complicated urinary tract infections (cUTI).

Perform susceptibility testing of multi-drug and extensively drug resistant aerobic Gram-negative organisms using Cefiderocol Antimicrobial Susceptibility Discs. Perform susceptibility testing of multi-drug and extensively drug resistant aerobic Gram-negative organisms using Cefiderocol Antimicrobial Susceptibility Discs. Features include:

  •  Meets EUCAST and CLSI recommended breakpoints
  • Produces quality results in line with the rest of the Oxoid disc portfolio as demonstrated over 3 EUCAST disc quality studies
  • Easy-to-use: shatterproof spring-loaded cartridges, easy-to-read labelling, and end-of-cartridge indicators.
  • Long-term stability ensured: each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%.
  • Accurate and reproducible: manufactured under stringent quality standards including BS EN ISO 9001:2008, ISO 13485:2003 and FDA 21 CFR part 820, with comprehensive quality control testing
  • Trusted supplier: over 35 years of experience in disc development and manufacture

Disc diffusion not only categorizes resistant, intermediate, and sensitive organisms through qualitative results but also provides a visual indication of the following:

  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics


Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing. 

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Cefiderocol
Concentration 30 μg
Description Cefiderocol Antimicrobial Susceptibility Discs
Antibiotic Code FDC30
Product Type Antimicrobial Susceptibility Discs
Target Organism Class Multidrug-resistant, Gram-negative Organisms, Extensively Drug-resistant (XDR), Gram-negative Organisms
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 250 Discs/Box
Quantity 50 Discs/Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.