Learn More
U.S. Pharmacopeia Cefixime, 125110-14-7, MFCD03788802, 500mg
Catalog No. NC9022198
Encompass
Molecular formula C16H15N5O7S2 . 3H2O, Molecular weight 507.5, Melting Point 424.4 - 437 °F (218 - 225 °C)
Catalog No. NC9022198 Supplier U.S. Pharmacopeia Supplier No. 1097658
Molecular formula C16H15N5O7S2 . 3H2O, Molecular weight 507.5, Melting Point 424.4 - 437 °F (218 - 225 °C)