Learn More
Alkali Scientific Cefixime Trihydrate, 100 Milligrams
SDP
Catalog No. 5000659800
Encompass_Preferred
Cefixime Trihydrate (100 mg) is a third-generation cephalosporin antibiotic used to treat a variety of bacterial infections. The powder form is specifically formulated for antibacterial use and works by inhibiting bacterial cell wall synthesis, resulting in bacterial cell death. Cefixime is highly effective against a range of bacterial infections, including respiratory tract infections, urinary tract infections, and certain types of ear infections. This 100 mg quantity is ideal for laboratory use and clinical preparations, offering a precise dosage for therapeutic purposes. It is an essential antibiotic in both medical and research settings for effective bacterial infection management.
Catalog No. 50-006-59800 Supplier Alkali Scientific Supplier No. EFI003CP
Cefixime Trihydrate (100 mg) is a third-generation cephalosporin antibiotic used to treat a variety of bacterial infections. The powder form is specifically formulated for antibacterial use and works by inhibiting bacterial cell wall synthesis, resulting in bacterial cell death. Cefixime is highly effective against a range of bacterial infections, including respiratory tract infections, urinary tract infections, and certain types of ear infections. This 100 mg quantity is ideal for laboratory use and clinical preparations, offering a precise dosage for therapeutic purposes. It is an essential antibiotic in both medical and research settings for effective bacterial infection management.