Learn More
Alkali Scientific Cefixime Trihydrate, 5 G
SDP
Catalog No. 5000659879
Encompass_Preferred
Cefixime Trihydrate, 5G, is an antibiotic used in laboratory research to evaluate antibacterial properties in microbial cultures. Cefixime is a third-generation cephalosporin that inhibits bacterial cell wall synthesis, rendering it effective against a wide range of Gram-positive and Gram-negative bacteria. The 5g volume allows for larger-scale experiments and testing, making it suitable for batch cultures in microbiological research. It is used in pharmaceutical studies, including the development and testing of new antibiotics, and is integral for investigating the effectiveness of antibacterial agents in treating infections. This formulation is specifically designed for research purposes and is not for clinical use.
Catalog No. 50-006-59879 Supplier Alkali Scientific Supplier No. EFI002CP
Cefixime Trihydrate, 5G, is an antibiotic used in laboratory research to evaluate antibacterial properties in microbial cultures. Cefixime is a third-generation cephalosporin that inhibits bacterial cell wall synthesis, rendering it effective against a wide range of Gram-positive and Gram-negative bacteria. The 5g volume allows for larger-scale experiments and testing, making it suitable for batch cultures in microbiological research. It is used in pharmaceutical studies, including the development and testing of new antibiotics, and is integral for investigating the effectiveness of antibacterial agents in treating infections. This formulation is specifically designed for research purposes and is not for clinical use.