Learn More
TARGETMOL CHEMICALS INC Cefmenoxime hydrochloride
SDP
Catalog No. 502879511
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cefmenoxime hydrochloride (Cefmenoxime hemihydrochloride) is a cephalosporin antibiotic that is administered intravenously or intramuscularly. It is active against most common gram-positive and gram-negative microorganisms, is a potent inhibitor of Enterobacteriaceae, and is highly resistant to hydrolysis by beta-lactamases. The drug has a high rate of efficacy in many types of infection and to date, no severe side effects have been noted. Purity 98.41%
Catalog No. 50-287-9511 Supplier TARGETMOL CHEMICALS INC Supplier No. T1190200MG
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cefmenoxime hydrochloride (Cefmenoxime hemihydrochloride) is a cephalosporin antibiotic that is administered intravenously or intramuscularly. It is active against most common gram-positive and gram-negative microorganisms, is a potent inhibitor of Enterobacteriaceae, and is highly resistant to hydrolysis by beta-lactamases. The drug has a high rate of efficacy in many types of infection and to date, no severe side effects have been noted. Purity 98.41%