Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cefmenoxime (hydrochloride) | 75738-58-8 | 99.3% | 1 G
SDP

Catalog No. 5000409420
Encompass_Preferred

Cefmenoxime hydrochloride is a new semisynthetic cephalosporin antibiotic, effective against a wide range of gram-positive and gram-negative bacteria.

  • Molecular weight: 529.79
  • Chemical formula: C16H17.5Cl0.5N9O5S3
  • Appearance: White to yellow solid
  • Storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in DMSO (65 mg/mL), insoluble in water
  • Inhibits at least 90% of tested strains (MIC90) from 0.06 to 8 μg/mL for Enterobacteriaceae
  • Effective against Haemophilus influenzae, Neisseria gonorrhoeae, and Neisseria meningitidis

Catalog No. 50-004-09420 Supplier Medchemexpress LLC Supplier No. HYB08751G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefmenoxime hydrochloride is a new semisynthetic cephalosporin antibiotic, effective against a wide range of gram-positive and gram-negative bacteria.

  • Molecular weight: 529.79
  • Chemical formula: C16H17.5Cl0.5N9O5S3
  • Appearance: White to yellow solid
  • Storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in DMSO (65 mg/mL), insoluble in water
  • Inhibits at least 90% of tested strains (MIC90) from 0.06 to 8 μg/mL for Enterobacteriaceae
  • Effective against Haemophilus influenzae, Neisseria gonorrhoeae, and Neisseria meningitidis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.