Learn More
Medchemexpress LLC Cefmenoxime hydrochloride | 75738-58-8 | 99.3% | 10 MM 1 ML
SDP
Catalog No. 5000407609
Encompass_Preferred
Cefmenoxime hydrochloride is a semisynthetic cephalosporin antibiotic. It demonstrates broad-spectrum antibacterial activity against both gram-positive and gram-negative bacteria.
  • Targets β-lactam
  • Inhibits 90% of tested strains (MIC90) from 0.06 to 8 μg/mL for Enterobacteriaceae
  • Effective against *Streptococcus pneumoniae*, *Streptococcus pyogenes*, and *S. aureus*
  • Active against *Haemophilus influenzae*, *Neisseria gonorrhoeae*, and *Neisseria meningitidis*
  • Improves survival rate in mice infected with lung bacteria
Catalog No. 50-004-07609 Supplier Medchemexpress LLC Supplier No. HYB087510MM/1ML
Cefmenoxime hydrochloride is a semisynthetic cephalosporin antibiotic. It demonstrates broad-spectrum antibacterial activity against both gram-positive and gram-negative bacteria.
  • Targets β-lactam
  • Inhibits 90% of tested strains (MIC90) from 0.06 to 8 μg/mL for Enterobacteriaceae
  • Effective against *Streptococcus pneumoniae*, *Streptococcus pyogenes*, and *S. aureus*
  • Active against *Haemophilus influenzae*, *Neisseria gonorrhoeae*, and *Neisseria meningitidis*
  • Improves survival rate in mice infected with lung bacteria