Learn More
Cayman Chemical Cefmetazole, Purity: ≥98%, CAS: 56796-20-4, 10 mg
SDP
Catalog No. 502989054
Encompass_Preferred
A cephamycin antibiotic; active against E. coli, P. mirabilis, H. influenzae, N. gonorrhoeae, S. pneumoniae, and S. pyogenes (MIC50s = 0.25-2 μg/ml); decreases the lesion size and the number of ulcerative lesions in a rabbit model of active syphilis at 20 and 40 mg/kg per day
Catalog No. 50-298-9054 Supplier Cayman Chemical Supplier No. 3026910 MG
A cephamycin antibiotic; active against E. coli, P. mirabilis, H. influenzae, N. gonorrhoeae, S. pneumoniae, and S. pyogenes (MIC50s = 0.25-2 μg/ml); decreases the lesion size and the number of ulcerative lesions in a rabbit model of active syphilis at 20 and 40 mg/kg per day