Learn More
Selleck Chemical LLC Cefmetazole sodium S5351-25mg
SDP
Catalog No. 503109967
Encompass_Preferred
Cefmetazole sodium is a semisynthetic cephamycin antibiotic with a broad spectrum of activity against both gram-positive and gram-negative microorganisms
Catalog No. 50-310-9967 Supplier Selleck Chemical LLC Supplier No. S535125MG
Add to Cart
Add to Cart
Cefmetazole sodium is a semisynthetic cephamycin antibiotic with a broad spectrum of activity against both gram-positive and gram-negative microorganisms