Learn More
Medchemexpress LLC Cefmetazole sodium | 56796-39-5 | 99.5% | 1 ML
SDP
Catalog No. 5000413116
Encompass_Preferred
Cefmetazole sodium is a semisynthetic cephamycin antibiotic that exhibits broad-spectrum antibacterial activity against gram-positive, gram-negative, and anaerobic bacteria. It functions by binding to penicillin binding proteins (PBPs), which interferes with the biosynthesis of bacterial cell walls. This product is intended for research purposes related to gynecologic, intraabdominal, urinary tract, respiratory tract, and skin and soft tissue infections.
  • Semisynthetic cephamycin antibiotic.
  • Broad-spectrum antibacterial activity.
  • Effective against gram-positive, gram-negative, and anaerobic bacteria.
  • Interferes with bacterial cell wall biosynthesis by binding to penicillin binding proteins (PBPs).
  • Used for research into gynecologic, intraabdominal, urinary tract, respiratory tract, and skin and soft tissue infections.
Catalog No. 50-004-13116 Supplier Medchemexpress LLC Supplier No. HYB125710MM/1ML
Cefmetazole sodium is a semisynthetic cephamycin antibiotic that exhibits broad-spectrum antibacterial activity against gram-positive, gram-negative, and anaerobic bacteria. It functions by binding to penicillin binding proteins (PBPs), which interferes with the biosynthesis of bacterial cell walls. This product is intended for research purposes related to gynecologic, intraabdominal, urinary tract, respiratory tract, and skin and soft tissue infections.
  • Semisynthetic cephamycin antibiotic.
  • Broad-spectrum antibacterial activity.
  • Effective against gram-positive, gram-negative, and anaerobic bacteria.
  • Interferes with bacterial cell wall biosynthesis by binding to penicillin binding proteins (PBPs).
  • Used for research into gynecologic, intraabdominal, urinary tract, respiratory tract, and skin and soft tissue infections.