Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefmetazole Sodium, G-Biosciences
SCI_ED

Catalog No. S774821
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S774821 1 g
S777032 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S774821 Supplier G-Biosciences Supplier No. RC925
Only null left
Add to Cart
Add to Cart

Cefmetazole is a semisynthetic second generation cephalosporin antibiotic derived from cephamycin. It is effective against a variety of gram-negative and gram-positive bacteria, exhibiting more efficacy against many gram-negative enteric bacteria than many first generation cephalosporins.

Cefmetazole sodium works by interfering with bacterial cell wall synthesis, ultimately leading to cell death.

Chemical Identifiers

Molecular Weight (g/mol) 493.52 g/mol

Specifications

Quantity 1 g
Percent Purity >92%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.