Learn More
Medchemexpress LLC HY-128932 25mg Medchemexpress, Cefminox (sodium) CAS:75498-96-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12893225MG
Encompass_Preferred
Medchemexpress, HY-128932 25mg Cefminox (sodium) CAS:75498-96-3 Cefminox sodium (MT-141) is a semisynthetic cephamycin, which exhibits a broad spectrum of antibacterial activity. Cefminox sodium (MT-141) also acts as a dual agonist of prostacyclin receptor (IP) and PPARγ, upregulates cAMP production and PTEN expression and inhibits Akt/mTOR signaling. Cefminox sodium (MT-141) also prevents pulmonary arterial hypertension. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1192