Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefminox Sodium Heptahydrate, G-Biosciences
SCI_ED

Catalog No. S777074
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S777074 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S777074 Supplier G-Biosciences Supplier No. RC927
Add to Cart
Add to Cart

Cefminox sodium is a second generation Beta-lactam cephalosporin antibiotic primarily used to treat bacterial infections.

cefminox sodium has exhibited a high degree of activity towards gram-negative and anaerobic bacteria. Cefminox has also been used to observe the effects of antibiotics on immunosuppressor and human serum albumin complexes.

Chemical Identifiers

CAS 1351813-81-4
Molecular Weight (g/mol) 667.66 g/mol

Specifications

Physical Form Flakes or Powder
Quantity 1 g
Percent Purity 98%
Grade Molecular Biology
Chemical Name or Material Cefminox Sodium Heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.