Learn More
Medchemexpress LLC Cefodizime sodium | 86329-79-5 | 99.9% | 628.63 | 100 MG
SDP
Catalog No. 5000346657
Encompass_Preferred
Cefodizime sodium is a third generation cephalosporin antibiotic with a broad spectrum of antibacterial activity. Cefodizime sodium has no renal toxic effect, good tolerance and immune regulation activity, and can be used for the research of severe infections of the respiratory and urinary tracts.
  • Broad spectrum of antibacterial activity
  • No renal toxic effect
  • Good tolerance
  • Immune regulation activity
  • Used for the research of severe infections of the respiratory and urinary tracts
Catalog No. 50-003-46657 Supplier Medchemexpress LLC Supplier No. HY108402A10MG
Cefodizime sodium is a third generation cephalosporin antibiotic with a broad spectrum of antibacterial activity. Cefodizime sodium has no renal toxic effect, good tolerance and immune regulation activity, and can be used for the research of severe infections of the respiratory and urinary tracts.
  • Broad spectrum of antibacterial activity
  • No renal toxic effect
  • Good tolerance
  • Immune regulation activity
  • Used for the research of severe infections of the respiratory and urinary tracts