Learn More
Medchemexpress LLC Cefodizime sodium | 86329-79-5 | 99.9% | 628.63 | 50 MG
SDP
Catalog No. 5000347366
Encompass_Preferred
Cefodizime sodium is a third-generation cephalosporin antibiotic with a broad spectrum of antibacterial activity. It has no renal toxic effect, good tolerance, and immune regulation activity. It can be used for the research of severe infections of the respiratory and urinary tracts.
  • Broad spectrum of antibacterial activity.
  • No renal toxic effect.
  • Good tolerance.
  • Immune regulation activity.
  • Suitable for research of severe respiratory and urinary tract infections.
  • Bactericidal antibiotic with high affinity for penicillin-binding proteins lA/B, 2, and 3 of E. coli.
  • Continues to show pronounced bactericidal activity for at least 48 hours after a single injection in studies.
  • Achieves complete bacterial clearance from the lung within 48 hours in 50% of mice in experimental studies.
Catalog No. 50-003-47366 Supplier Medchemexpress LLC Supplier No. HY108402A100MG
Cefodizime sodium is a third-generation cephalosporin antibiotic with a broad spectrum of antibacterial activity. It has no renal toxic effect, good tolerance, and immune regulation activity. It can be used for the research of severe infections of the respiratory and urinary tracts.
  • Broad spectrum of antibacterial activity.
  • No renal toxic effect.
  • Good tolerance.
  • Immune regulation activity.
  • Suitable for research of severe respiratory and urinary tract infections.
  • Bactericidal antibiotic with high affinity for penicillin-binding proteins lA/B, 2, and 3 of E. coli.
  • Continues to show pronounced bactericidal activity for at least 48 hours after a single injection in studies.
  • Achieves complete bacterial clearance from the lung within 48 hours in 50% of mice in experimental studies.