Learn More
Medchemexpress LLC Cefonicid sodium | 61270-78-8 | 95.0% | 1 ML
SDP
Catalog No. 5000412545
Encompass_Preferred
Cefonicid sodium is a long-acting cephalosporin antibiotic. It also acts as a noncompetitive class I β-lactamase inhibitor with a Kᵢ value of 0.8 μM. It shows broad-spectrum antimicrobial activity and is effective against a variety of Gram-positive and Gram-negative bacteria. It can be used for research on infections.
  • Long-acting cephalosporin antibiotic
  • Noncompetitive class I β-lactamase inhibitor
  • Exhibits broad-spectrum antimicrobial activity
  • Effective against Gram-positive and Gram-negative bacteria
  • Suitable for research on infections
Catalog No. 50-004-12545 Supplier Medchemexpress LLC Supplier No. HYB130010MM/1ML
Cefonicid sodium is a long-acting cephalosporin antibiotic. It also acts as a noncompetitive class I β-lactamase inhibitor with a Kᵢ value of 0.8 μM. It shows broad-spectrum antimicrobial activity and is effective against a variety of Gram-positive and Gram-negative bacteria. It can be used for research on infections.
  • Long-acting cephalosporin antibiotic
  • Noncompetitive class I β-lactamase inhibitor
  • Exhibits broad-spectrum antimicrobial activity
  • Effective against Gram-positive and Gram-negative bacteria
  • Suitable for research on infections