Learn More
Apexbio Technology LLC Cefonicid (sodium salt) 61270-78-8 1g
SDP
Catalog No. 5000091241
Encompass_Preferred
Cefonicid sodium salt (CAS 61270-78-8) is a second-generation cephalosporin antibiotic targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby exerting bactericidal effects against susceptible bacteria Cefonicid sodium salt acts through inhibition of bacterial cell wall synthesis by binding to PBPs In vitro studies indicate potent inhibition of pathogens such as Neisseria gonorrhoeae with minimal inhibitory concentration (MIC) values typically 2 g/mL Based on these pharmacological properties cefonicid sodium salt holds research potential for studying and treating urinary tract infections lower respiratory tract infections soft tissue infections osteomyelitis and uncomplicated gonorrhea
Catalog No. 50-000-91241 Supplier Apexbio Technology LLC Supplier No. C38121000
Cefonicid sodium salt (CAS 61270-78-8) is a second-generation cephalosporin antibiotic targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby exerting bactericidal effects against susceptible bacteria Cefonicid sodium salt acts through inhibition of bacterial cell wall synthesis by binding to PBPs In vitro studies indicate potent inhibition of pathogens such as Neisseria gonorrhoeae with minimal inhibitory concentration (MIC) values typically 2 g/mL Based on these pharmacological properties cefonicid sodium salt holds research potential for studying and treating urinary tract infections lower respiratory tract infections soft tissue infections osteomyelitis and uncomplicated gonorrhea