Learn More
Apexbio Technology LLC Cefoperazone, 1g. Cas: 62893-19-0 MFCD: MFCD00865067. Third-generation cephalosporin antibiotic
SDP
Catalog No. 501905135
Encompass_Preferred
Cefoperazone is a cephalosporin antibiotic which inhibits rMrp2-mediated [3H]E217G uptake with an IC50 of 199 M [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5135 Supplier Apexbio Technology LLC Supplier No. A83921000
Cefoperazone is a cephalosporin antibiotic which inhibits rMrp2-mediated [3H]E217G uptake with an IC50 of 199 M [1]. Other sizes are available. Please inqury us for quote.