Learn More
Research Products International Corp Cefoperazone, 500 MG
SDP
Catalog No. 501978371
Encompass_Preferred
Cefoperazone Sodium Salt is a third-generation cephalosporin antibiotic. Cefoperazone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs). This disruption interferes with the final stages of peptidoglycan synthesis in the bacterial cell wall, leading to structural instability and eventual cell lysis. 
Cefoperazone is effective against a broad range of bacteria, both Gram-positive and Gram-negative. Cefoperazone Sodium Salt is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion allows for the specific growth of bacteria lacking resistance to cefoperazone.
Catalog No. 50-197-8371 Supplier Research Products International Corp Supplier No. C576000.5
Cefoperazone Sodium Salt is a third-generation cephalosporin antibiotic. Cefoperazone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs). This disruption interferes with the final stages of peptidoglycan synthesis in the bacterial cell wall, leading to structural instability and eventual cell lysis. 
Cefoperazone is effective against a broad range of bacteria, both Gram-positive and Gram-negative. Cefoperazone Sodium Salt is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion allows for the specific growth of bacteria lacking resistance to cefoperazone.