Learn More
Medchemexpress LLC Cefoperazone-d5 | 2410425-70-4 | 98.6% | 100 UG
SDP
Catalog No. 5000405714
Encompass_Preferred
Cefoperazone-d5 is deuterium labeled Cefoperazone. Cefoperazone, a semisynthetic cephalosporin, has a broad spectrum of antibacterial activity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration is noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-05714 Supplier Medchemexpress LLC Supplier No. HYB0210S100UG
Cefoperazone-d5 is deuterium labeled Cefoperazone. Cefoperazone, a semisynthetic cephalosporin, has a broad spectrum of antibacterial activity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration is noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.