Learn More
Apexbio Technology LLC Cefoperazone (sodium salt) 62893-20-3 10g
SDP
Catalog No. 5000091335
Encompass_Preferred
Cefoperazone sodium salt (CAS 62893-20-3) is a semi-synthetic cephalosporin antibiotic targeting penicillin-binding proteins It is designed to inhibit bacterial cell wall synthesis thereby compromising cell wall stability and inducing bacterial cell lysis Cefoperazone sodium salt exerts its biological activity primarily through inhibition of bacterial cell wall synthesis In in vitro studies cefoperazone sodium salt demonstrates broad-spectrum antibacterial activity with MIC50 values ranging from 0 004 0 06 g/ml against Neisseria gonorrhoeae isolates Based on these pharmacological properties cefoperazone sodium salt holds research potential in experimental infection models to investigate bacterial susceptibility resistance mechanisms and pharmacokinetic properties in clinical microbiology and infectious diseases
Catalog No. 50-000-91335 Supplier Apexbio Technology LLC Supplier No. C391310000
Cefoperazone sodium salt (CAS 62893-20-3) is a semi-synthetic cephalosporin antibiotic targeting penicillin-binding proteins It is designed to inhibit bacterial cell wall synthesis thereby compromising cell wall stability and inducing bacterial cell lysis Cefoperazone sodium salt exerts its biological activity primarily through inhibition of bacterial cell wall synthesis In in vitro studies cefoperazone sodium salt demonstrates broad-spectrum antibacterial activity with MIC50 values ranging from 0 004 0 06 g/ml against Neisseria gonorrhoeae isolates Based on these pharmacological properties cefoperazone sodium salt holds research potential in experimental infection models to investigate bacterial susceptibility resistance mechanisms and pharmacokinetic properties in clinical microbiology and infectious diseases