Learn More
Medchemexpress LLC Cefoperazone sodium salt | 62893-20-3 | 99.55% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0210A10MM/1ML
Encompass_Preferred
Cefoperazone sodium salt (CP 52640-2) is a semisynthetic cephalosporin with a broad spectrum of antibacterial activity.
  • Semisynthetic cephalosporin.
  • Demonstrates broad spectrum antibacterial activity.
  • Protects granulocytopenic mice from acute *Pseudomonas aeruginosa* pneumonia in aerosol treatment.
Catalog No. 50-004-05775