Learn More
Medchemexpress LLC Cefoperazone sodium salt | 62893-20-3 | 99.55% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0210A5G
Encompass_Preferred
Cefoperazone sodium salt is a semisynthetic cephalosporin with broad-spectrum antibacterial activity, intended for research use only. It appears as a white to off-white solid and is soluble in both DMSO and water.
  • Exhibits broad-spectrum antibacterial activity
  • Intended for research use only
  • Soluble in DMSO and water
  • Protects against acute Pseudomonas aeruginosa pneumonia in granulocytopenic mice
Catalog No. 50-004-05400