Learn More
Medchemexpress LLC Cefoperazone (sodium salt) (Standard) | 62893-20-3 | 99.1% | 250 MG
SDP
Catalog No. 5000405604
Encompass_Preferred
Cefoperazone (sodium salt) (Standard) is the analytical standard of Cefoperazone (sodium salt). This product is intended for research and analytical applications. Cefoperazone sodium salt (CP 52640-2), a semisynthetic cephalosporin, has a broad spectrum of antibacterial activity.
  • Analytical standard for Cefoperazone (sodium salt)
  • Intended for research and analytical applications
  • Semisynthetic cephalosporin with broad-spectrum antibacterial activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Store at 4°C, sealed and dry, away from light and moisture
  • Shipping at room temperature for short durations (less than 2 weeks)
Catalog No. 50-004-05604 Supplier Medchemexpress LLC Supplier No. 250MGHYB0210AR
Cefoperazone (sodium salt) (Standard) is the analytical standard of Cefoperazone (sodium salt). This product is intended for research and analytical applications. Cefoperazone sodium salt (CP 52640-2), a semisynthetic cephalosporin, has a broad spectrum of antibacterial activity.
  • Analytical standard for Cefoperazone (sodium salt)
  • Intended for research and analytical applications
  • Semisynthetic cephalosporin with broad-spectrum antibacterial activity
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Store at 4°C, sealed and dry, away from light and moisture
  • Shipping at room temperature for short durations (less than 2 weeks)