Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefoperazone Sodium, USP Grade, G-Biosciences
SCI_ED

Catalog No. S891101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S891101 1 g
S000009 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S891101 Supplier G-Biosciences Supplier No. RC928
Add to Cart
Add to Cart

Cefoperazone is a third generation cephalosporin antibiotic primarily targeting P. aeruginosa and other gram-negative bacteria.

It has also been used to study penicillin binding proteins (PBPs) and is a Beta-lactamase inhibitor.

Chemical Identifiers

Molecular Weight (g/mol) 667.65 g/mol

Specifications

Boiling Point 200 to 202°C
Quantity 1 g
Percent Purity >98%
Grade USP
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.