Learn More
Medchemexpress LLC HY-B0186B 10mg Medchemexpress, Cefoselis (sulfate) CAS:122841-12-7 Purity:>98%
SDP
Catalog No. 502082403
Encompass_Preferred
Medchemexpress, HY-B0186B 10mg Cefoselis (sulfate) CAS:122841-12-7 Cefoselis sulfate (FK-037), the fourth gen-eration of cephalosporin, is a β-lactam antibiotic. Cefoselis sulfate exhibits good activity against a wide range of Gram-positive and Gram-negative organisms. Cefoselis sulfate penetrates the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2403 Supplier Medchemexpress LLC Supplier No. HYB0186B10MG
Medchemexpress, HY-B0186B 10mg Cefoselis (sulfate) CAS:122841-12-7 Cefoselis sulfate (FK-037), the fourth gen-eration of cephalosporin, is a β-lactam antibiotic. Cefoselis sulfate exhibits good activity against a wide range of Gram-positive and Gram-negative organisms. Cefoselis sulfate penetrates the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.