Learn More
Apexbio Technology LLC Cefoselis Sulfate 122841-12-7 50mg
SDP
Catalog No. 5000089930
Encompass_Preferred
Cefoselis Sulfate (CAS 122841-12-7) is a beta-lactam antibiotic targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis by covalently binding PBPs thereby disrupting peptidoglycan cross-linking and leading to bacterial cell lysis Cefoselis Sulfate exerts its biological activity primarily through inhibition of bacterial cell wall biosynthesis In in vitro antimicrobial assays Cefoselis Sulfate demonstrates antibacterial activity with reported IC50 values varying depending on the bacterial strain and experimental conditions Based on these pharmacological properties Cefoselis Sulfate holds research potential in studies examining antimicrobial resistance patterns bacterial sensitivity profiles and bacterial resistance mechanisms
Catalog No. 50-000-89930 Supplier Apexbio Technology LLC Supplier No. B190650
Cefoselis Sulfate (CAS 122841-12-7) is a beta-lactam antibiotic targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis by covalently binding PBPs thereby disrupting peptidoglycan cross-linking and leading to bacterial cell lysis Cefoselis Sulfate exerts its biological activity primarily through inhibition of bacterial cell wall biosynthesis In in vitro antimicrobial assays Cefoselis Sulfate demonstrates antibacterial activity with reported IC50 values varying depending on the bacterial strain and experimental conditions Based on these pharmacological properties Cefoselis Sulfate holds research potential in studies examining antimicrobial resistance patterns bacterial sensitivity profiles and bacterial resistance mechanisms