Learn More
Medchemexpress LLC Cefoselis sulfate | 122841-12-7 | 99.4% | 100 MG
SDP
Catalog No. 5000407004
Encompass_Preferred
Cefoselis sulfate (FK-037) is a fourth-generation cephalosporin and a β-lactam antibiotic. It shows good activity against a wide range of Gram-positive and Gram-negative organisms and can penetrate the blood-brain barrier.
  • Is a fourth-generation cephalosporin
  • Functions as a β-lactam antibiotic
  • Exhibits good activity against a wide range of Gram-positive and Gram-negative organisms
  • Penetrates the blood-brain barrier
  • Activity is similar to that of cefpirome and cefepime, and generally superior to ceftazidime
  • Shows potent antibacterial activity against Hemophilus influenzae and Moraxella catarrhalis
  • Highly active against MSSA and MSCNS
Catalog No. 50-004-07004 Supplier Medchemexpress LLC Supplier No. HYB0186B100MG
Cefoselis sulfate (FK-037) is a fourth-generation cephalosporin and a β-lactam antibiotic. It shows good activity against a wide range of Gram-positive and Gram-negative organisms and can penetrate the blood-brain barrier.
  • Is a fourth-generation cephalosporin
  • Functions as a β-lactam antibiotic
  • Exhibits good activity against a wide range of Gram-positive and Gram-negative organisms
  • Penetrates the blood-brain barrier
  • Activity is similar to that of cefpirome and cefepime, and generally superior to ceftazidime
  • Shows potent antibacterial activity against Hemophilus influenzae and Moraxella catarrhalis
  • Highly active against MSSA and MSCNS