Learn More
TARGETMOL CHEMICALS INC Cefoselis Sulfate 25MG
SDP
Catalog No. 502877329
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 100 mg and bulk. Please contact Fisher for quotes. Cefoselis Sulfate is a widely used beta-lactam antibiotic. Purity 99.84%
Catalog No. 50-287-7329 Supplier TARGETMOL CHEMICALS INC Supplier No. T643825MG
Also available in 1 mL, 5 mg, 10 mg, 100 mg and bulk. Please contact Fisher for quotes. Cefoselis Sulfate is a widely used beta-lactam antibiotic. Purity 99.84%