Learn More
Medchemexpress LLC Cefoselis sulfate (FK-037) | 122841-12-7 | MFCD01940015 | 99.4% | 50 MG
SDP
Catalog No. 5000405798
Encompass_Preferred
Cefoselis sulfate (FK-037) is a fourth-generation cephalosporin and a β-lactam antibiotic known for its broad-spectrum activity. It is effective against a wide range of both Gram-positive and Gram-negative organisms. A key feature of this compound is its ability to penetrate the blood-brain barrier.
  • Demonstrates good activity against a broad spectrum of Gram-positive and Gram-negative organisms.
  • Ability to penetrate the blood-brain barrier.
  • Functions as a fourth-generation cephalosporin and β-lactam antibiotic.
  • High purity.
Catalog No. 50-004-05798 Supplier Medchemexpress LLC Supplier No. HYB0186B50MG
Cefoselis sulfate (FK-037) is a fourth-generation cephalosporin and a β-lactam antibiotic known for its broad-spectrum activity. It is effective against a wide range of both Gram-positive and Gram-negative organisms. A key feature of this compound is its ability to penetrate the blood-brain barrier.
  • Demonstrates good activity against a broad spectrum of Gram-positive and Gram-negative organisms.
  • Ability to penetrate the blood-brain barrier.
  • Functions as a fourth-generation cephalosporin and β-lactam antibiotic.
  • High purity.