Learn More
Selleck Chemical LLC Cefotaxime S4720-25mg
SDP
Catalog No. 503108438
Encompass_Preferred
Cefotaxime(HR-756) is a third-generation cephalosporin antibiotic and has broad spectrum activity against Gram positive and Gram negative bacteria
Catalog No. 50-310-8438 Supplier Selleck Chemical LLC Supplier No. S472025MG
Cefotaxime(HR-756) is a third-generation cephalosporin antibiotic and has broad spectrum activity against Gram positive and Gram negative bacteria