Learn More
Medchemexpress LLC Cefotaxime | 63527-52-6 | 98.7% | C16H17N5O7S2 | 500 MG
SDP
Catalog No. 5000432367
Encompass_Preferred
Cefotaxime is a β-lactamase stable cephalosporin and a third-generation cephalosporin antibiotic. It possesses broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria. This product is for research use only and not sold to patients.
  • Beta-lactamase stable cephalosporin
  • Third-generation cephalosporin antibiotic
  • Broad-spectrum antibiotic activity
  • Effective against Gram-positive and Gram-negative bacteria
  • Suitable for research use
Catalog No. 50-004-32367 Supplier Medchemexpress LLC Supplier No. HYA0088A500MG
Cefotaxime is a β-lactamase stable cephalosporin and a third-generation cephalosporin antibiotic. It possesses broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria. This product is for research use only and not sold to patients.
  • Beta-lactamase stable cephalosporin
  • Third-generation cephalosporin antibiotic
  • Broad-spectrum antibiotic activity
  • Effective against Gram-positive and Gram-negative bacteria
  • Suitable for research use