Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cefotaxime-d3 | 63527-52-6 | 98.0% | 458.48 g/mol | C16H14D3N5O7S2 | 1 MG
SDP

Catalog No. 5000283917
Encompass_Preferred

Cefotaxime-d3 is the deuterium-labeled isotopologue of the third-generation cephalosporin cefotaxime, supplied as a solid for use as an analytical internal standard and tracer in quantitative methods such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative assays.
  • High purity: 98.0%.
  • Chemical formula: C16H14D3N5O7S2.
  • Molecular weight: 458.48 g/mol.
  • Appearance: white to off-white solid.
  • Intended applications: tracer, calibration, and bioanalytical quantitation.
  • Available in 1 mg size; larger sizes available by quote.

Catalog No. 50-002-83917 Supplier Medchemexpress LLC Supplier No. HYW6541011MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefotaxime-d3 is the deuterium-labeled isotopologue of the third-generation cephalosporin cefotaxime, supplied as a solid for use as an analytical internal standard and tracer in quantitative methods such as NMR, GC-MS, and LC-MS.

  • Deuterium-labeled internal standard for quantitative assays.
  • High purity: 98.0%.
  • Chemical formula: C16H14D3N5O7S2.
  • Molecular weight: 458.48 g/mol.
  • Appearance: white to off-white solid.
  • Intended applications: tracer, calibration, and bioanalytical quantitation.
  • Available in 1 mg size; larger sizes available by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.