Learn More
Gold Biotechnology Inc Cefotaxime (Sodium), USP Grade 5 g
SDP
Catalog No. 501532755
Encompass_Preferred
Cefotaxime is a third generation, broad-spectrum cephalosporin antibiotic effective against various gram-negative and gram-positive bacteria. However, unlike many other cephalosporins, cefotaxime is not effective against
Catalog No. 50-153-2755 Supplier Gold Biotechnology Inc Supplier No. C1045
Cefotaxime is a third generation, broad-spectrum cephalosporin antibiotic effective against various gram-negative and gram-positive bacteria. However, unlike many other cephalosporins, cefotaxime is not effective against