Learn More
Medchemexpress LLC Cefotaxime sodium | 64485-93-4 | 99.1% | C16H16N5NaO7S2 | 1 ML
SDP
Catalog No. 5000432107
Encompass_Preferred
Cefotaxime sodium is a β-lactamase stable, third-generation cephalosporin antibiotic. It demonstrates broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria.
  • Exhibits MIC of 0.0625 mg/L for V. vulnificus CMCP6
  • More effective in clearing V. vulnificus in vivo when combined with ciprofloxacin
  • Lowers viable bacterial counts in liver when combined with ciprofloxacin
Catalog No. 50-004-32107 Supplier Medchemexpress LLC Supplier No. HYA008810MM/1ML
Cefotaxime sodium is a β-lactamase stable, third-generation cephalosporin antibiotic. It demonstrates broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria.
  • Exhibits MIC of 0.0625 mg/L for V. vulnificus CMCP6
  • More effective in clearing V. vulnificus in vivo when combined with ciprofloxacin
  • Lowers viable bacterial counts in liver when combined with ciprofloxacin