Learn More
Medchemexpress LLC Cefotaxime (sodium) | 64485-93-4 | 99.1% | C16H16N5NaO7S2 | 5 G
SDP
Catalog No. 5000432338
Encompass_Preferred
Cefotaxime sodium is a third-generation cephalosporin antibiotic that is stable against β-lactamase. It exhibits broad-spectrum antibiotic activity against a wide range of Gram-positive and Gram-negative bacteria. It is intended for research use only.
  • β-lactamase stable cephalosporin, third-generation cephalosporin antibiotic.
  • Possesses broad-spectrum antibiotic activity.
  • Effective against numerous Gram-positive and Gram-negative bacteria.
  • For research use only.
Catalog No. 50-004-32338 Supplier Medchemexpress LLC Supplier No. HYA00885G
Cefotaxime sodium is a third-generation cephalosporin antibiotic that is stable against β-lactamase. It exhibits broad-spectrum antibiotic activity against a wide range of Gram-positive and Gram-negative bacteria. It is intended for research use only.
  • β-lactamase stable cephalosporin, third-generation cephalosporin antibiotic.
  • Possesses broad-spectrum antibiotic activity.
  • Effective against numerous Gram-positive and Gram-negative bacteria.
  • For research use only.