Learn More
Medchemexpress LLC Cefotaxime sodium | 64485-93-4 | 99.1% | C16H16N5NaO7S2 | 500 MG
SDP
Catalog No. 5000432378
Encompass_Preferred
Cefotaxime sodium is a β-lactamase stable cephalosporin and a third-generation cephalosporin antibiotic. It demonstrates broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria.
  • Beta-lactamase stable cephalosporin
  • Third-generation cephalosporin antibiotic
  • Broad-spectrum antibiotic activity
  • Active against numerous Gram-positive and Gram-negative bacteria
  • Exhibits an MIC of 0.0625 mg/L for *V. vulnificus* CMCP6
Catalog No. 50-004-32378 Supplier Medchemexpress LLC Supplier No. HYA0088500MG
Cefotaxime sodium is a β-lactamase stable cephalosporin and a third-generation cephalosporin antibiotic. It demonstrates broad-spectrum antibiotic activity against numerous Gram-positive and Gram-negative bacteria.
  • Beta-lactamase stable cephalosporin
  • Third-generation cephalosporin antibiotic
  • Broad-spectrum antibiotic activity
  • Active against numerous Gram-positive and Gram-negative bacteria
  • Exhibits an MIC of 0.0625 mg/L for *V. vulnificus* CMCP6