Learn More
Bioworld Cefotaxime Sodium, 5 g
Catalog No. 50255713
Encompass_Preferred
°°Cefotaxime SodiumBactericidal activityInhibits cell wall synthesisIn vitro activity against a wide range of gram-positive and gram-negative organismsHas a high degree of stability in the presence of -lactamases, both penicillinases and cephalosporinases, of gram-negative and gram-positive bacteriaSynonym: Cephotaxime; Sym-methoxyimino Cephalosporin; ClaforanFor in vitro lab use only. Not for human use or consumption.RTECS Number: XI0250000
Catalog No. 50-255-713 Supplier Bioworld Supplier No. 403100081
°°Cefotaxime SodiumBactericidal activityInhibits cell wall synthesisIn vitro activity against a wide range of gram-positive and gram-negative organismsHas a high degree of stability in the presence of -lactamases, both penicillinases and cephalosporinases, of gram-negative and gram-positive bacteriaSynonym: Cephotaxime; Sym-methoxyimino Cephalosporin; ClaforanFor in vitro lab use only. Not for human use or consumption.RTECS Number: XI0250000