Learn More
Cayman Chemical Cefotaxime (sodium salt), Purity: ≥98%, CAS: 64485-93-4, 500 mg
SDP
Catalog No. 502972196
Encompass_Preferred
A cephalosporin antibiotic with potent in vitro activity against streptococcal species, including S. aureus (IC50 = 2 µg/ml) and penicillin-resistant S. pneumonia and also effective against a wide range of other β-lactamase-producing bacterial species (MIC90s = 0.01 - 130 µg/ml)
Catalog No. 50-297-2196 Supplier Cayman Chemical Supplier No. 16040500 MG
A cephalosporin antibiotic with potent in vitro activity against streptococcal species, including S. aureus (IC50 = 2 µg/ml) and penicillin-resistant S. pneumonia and also effective against a wide range of other β-lactamase-producing bacterial species (MIC90s = 0.01 - 130 µg/ml)