Learn More
Apexbio Technology LLC CEFOTETAN-10MG
SDP
Catalog No. 5000568889
Encompass_Preferred
Cefotetan (CAS No 69712-56-7) is a cephamycin antibiotic primarily used in the field of microbiology and biomedical research Originating from bacterial sources it acts by inhibiting bacterial cell wall synthesis targeting penicillin-binding proteins and disrupting peptidoglycan formation It demonstrates significant in vitro activity often observed in the nanomolar to low micromolar range against a broad spectrum of Gram-negative and Gram-positive bacteria Cefotetan is commonly applied in cell models and animal studies to evaluate antibacterial efficacy and resistance patterns It is also utilized in drug screening assays to identify potential antibiotic combinations or resistance mechanisms Experimental concentrations typically depend on the specific research design Cefotetan s role as a model compound in understanding bacterial cell wall synthesis and resistance pathways makes it a valuable tool in antibiotic research
Catalog No. 50-005-68889 Supplier Apexbio Technology LLC Supplier No. BA106310
Cefotetan (CAS No 69712-56-7) is a cephamycin antibiotic primarily used in the field of microbiology and biomedical research Originating from bacterial sources it acts by inhibiting bacterial cell wall synthesis targeting penicillin-binding proteins and disrupting peptidoglycan formation It demonstrates significant in vitro activity often observed in the nanomolar to low micromolar range against a broad spectrum of Gram-negative and Gram-positive bacteria Cefotetan is commonly applied in cell models and animal studies to evaluate antibacterial efficacy and resistance patterns It is also utilized in drug screening assays to identify potential antibiotic combinations or resistance mechanisms Experimental concentrations typically depend on the specific research design Cefotetan s role as a model compound in understanding bacterial cell wall synthesis and resistance pathways makes it a valuable tool in antibiotic research