Learn More
Medchemexpress LLC Cefotetan | 69712-56-7 | 25 MG
SDP
Catalog No. 5000421779
Encompass_Preferred
Cefotetan targets human Raf1 kinase inhibitor protein (hRKIP), enhancing ERK phosphorylation by relieving hRKIP's inhibition of the Ras/Raf1/MEK/ERK signaling pathway. It is used to study diseases associated with dysregulated Ras/Raf1/MEK/ERK signaling. Additionally, Cefotetan is a broad-spectrum antibacterial agent that disrupts cell wall synthesis by binding to bacterial penicillin-binding proteins (PBPs), useful for studying bacterial infections in bone, skin, urinary tract, and lower respiratory tract.
  • Targets human Raf1 kinase inhibitor protein (hRKIP)
  • Enhances ERK phosphorylation
  • Broad-spectrum antibacterial agent
  • Disrupts bacterial cell wall synthesis via penicillin-binding proteins
  • Active against gram-positive and gram-negative bacteria
  • Highly stable against beta-lactamases
  • Appearance: Solid, white to off-white
Catalog No. 50-004-21779 Supplier Medchemexpress LLC Supplier No. 25MGHYN6670
Cefotetan targets human Raf1 kinase inhibitor protein (hRKIP), enhancing ERK phosphorylation by relieving hRKIP's inhibition of the Ras/Raf1/MEK/ERK signaling pathway. It is used to study diseases associated with dysregulated Ras/Raf1/MEK/ERK signaling. Additionally, Cefotetan is a broad-spectrum antibacterial agent that disrupts cell wall synthesis by binding to bacterial penicillin-binding proteins (PBPs), useful for studying bacterial infections in bone, skin, urinary tract, and lower respiratory tract.
  • Targets human Raf1 kinase inhibitor protein (hRKIP)
  • Enhances ERK phosphorylation
  • Broad-spectrum antibacterial agent
  • Disrupts bacterial cell wall synthesis via penicillin-binding proteins
  • Active against gram-positive and gram-negative bacteria
  • Highly stable against beta-lactamases
  • Appearance: Solid, white to off-white