Learn More
Medchemexpress LLC Cefotetan | 69712-56-7 | 500 MG
SDP
Catalog No. 5000417921
Encompass_Preferred
Cefotetan is a binding agent that targets human Raf1 kinase inhibitor protein (hRKIP), enhancing ERK phosphorylation by relieving hRKIP's inhibition of the Ras/Raf1/MEK/ERK signaling pathway. It is also a broad-spectrum antibacterial agent that disrupts cell wall synthesis by binding to bacterial penicillin-binding proteins (PBPs). Cefotetan is used to study diseases associated with dysregulated Ras/Raf1/MEK/ERK signaling pathways and bacterial infections.
  • Targets human Raf1 kinase inhibitor protein (hRKIP).
  • Enhances ERK phosphorylation.
  • Broad-spectrum antibacterial activity.
  • Disrupts bacterial cell wall synthesis.
  • Effective against Gram-positive and Gram-negative bacteria.
  • Highly stable against β-lactamases.
Catalog No. 50-004-17921 Supplier Medchemexpress LLC Supplier No. HYN6670500MG
Cefotetan is a binding agent that targets human Raf1 kinase inhibitor protein (hRKIP), enhancing ERK phosphorylation by relieving hRKIP's inhibition of the Ras/Raf1/MEK/ERK signaling pathway. It is also a broad-spectrum antibacterial agent that disrupts cell wall synthesis by binding to bacterial penicillin-binding proteins (PBPs). Cefotetan is used to study diseases associated with dysregulated Ras/Raf1/MEK/ERK signaling pathways and bacterial infections.
  • Targets human Raf1 kinase inhibitor protein (hRKIP).
  • Enhances ERK phosphorylation.
  • Broad-spectrum antibacterial activity.
  • Disrupts bacterial cell wall synthesis.
  • Effective against Gram-positive and Gram-negative bacteria.
  • Highly stable against β-lactamases.