Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefotetan Disodium, G-Biosciences
SCI_ED

Catalog No. S000010
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000010 1 g
S902843 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000010 Supplier G-Biosciences Supplier No. RC933
Add to Cart
Add to Cart

Cefotetan disodium is a semi-synthetic Beta-lactam antibiotic that is classified as a second generation cephalosporin.

Cefotetan disodium effects a broad range of gram positive and gram negative bacteria. It has also displayed a significant degree of bactericidal activity towards aerobic and anaerobic bacteria.

Chemical Identifiers

Molecular Weight (g/mol) 619.58 g/mol

Specifications

Boiling Point 798°C
Quantity 1 g
Percent Purity >95%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.