Learn More
Cayman Chemical Cefotetan (sodium salt), Purity: ≥95%, CAS: 74356-00-6, 100 mg
SDP
Catalog No. 502994087
Encompass_Preferred
A cephamycin antibiotic; active against Gram-positive and Gram-negative bacteria (MICs = 0.03-32 mg/ml); reduces mortality by 50% in a rabbit model of fecal peritonitis (25 mg/kg twice daily)
Catalog No. 50-299-4087 Supplier Cayman Chemical Supplier No. 23624100 MG
A cephamycin antibiotic; active against Gram-positive and Gram-negative bacteria (MICs = 0.03-32 mg/ml); reduces mortality by 50% in a rabbit model of fecal peritonitis (25 mg/kg twice daily)