Learn More
Cayman Chemical CefotetnsodIum salt 50mg
SDP
Catalog No. 502994162
Encompass_Preferred
A cephamycin antibiotic; active against Gram-positive and Gram-negative bacteria (MICs = 0.03-32 mg/ml); reduces mortality by 50% in a rabbit model of fecal peritonitis (25 mg/kg twice daily)
Catalog No. 50-299-4162 Supplier Cayman Chemical Supplier No. 2362450 MG
A cephamycin antibiotic; active against Gram-positive and Gram-negative bacteria (MICs = 0.03-32 mg/ml); reduces mortality by 50% in a rabbit model of fecal peritonitis (25 mg/kg twice daily)