Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cefotiam hexetil hydrochloride | 95789-30-3 | MFCD00897074 | 95.1% | 768.76 | C27H39Cl2N9O7S3 | 10 MG
SDP

Catalog No. 5000298780
Encompass_Preferred

Cefotiam hexetil hydrochloride is the hydrochloride salt of cefotiam hexetil, an oral prodrug of the third-generation cephalosporin cefotiam. Intended for research and analytical use, the compound is used in prodrug, formulation, and pharmacology studies and is inactive as an antibacterial until converted to cefotiam.

  • Hydrochloride salt with reported purity 95.1%
  • Molecular formula C27H39Cl2N9O7S3 and molecular weight 768.76
  • Soluble in DMSO (250 mg/mL); ultrasonic agitation recommended
  • Appearance: solid, off-white to light yellow
  • Storage: store sealed at 4°C, protect from moisture and light
  • Supplied in small research sizes such as 10 mg

Catalog No. 50-002-98780 Supplier Medchemexpress LLC Supplier No. HYA0110A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefotiam hexetil hydrochloride is the hydrochloride salt of cefotiam hexetil, an oral prodrug of the third-generation cephalosporin cefotiam. Intended for research and analytical use, the compound is used in prodrug, formulation, and pharmacology studies and is inactive as an antibacterial until converted to cefotiam.

  • Hydrochloride salt with reported purity 95.1%
  • Molecular formula C27H39Cl2N9O7S3 and molecular weight 768.76
  • Soluble in DMSO (250 mg/mL); ultrasonic agitation recommended
  • Appearance: solid, off-white to light yellow
  • Storage: store sealed at 4°C, protect from moisture and light
  • Supplied in small research sizes such as 10 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.