Learn More
Medchemexpress LLC Cefotiam hexetil hydrochloride | 95789-30-3 | MFCD00897074 | 95.1% | C27H39Cl2N9O7S3 | 50 MG
SDP
Catalog No. 5000432653
Encompass_Preferred
Cefotiam hexetil hydrochloride is an oral third-generation cephalosporin. It functions as a proagent of cefotiam, an antibiotic, though it does not possess anti-bacterial properties itself. This product is intended for research use only and is not sold to patients.
  • Functions as a proagent of cefotiam
  • Oral third-generation cephalosporin
  • Cefotiam is known as an antibiotic
  • Suitable for research use only
  • Off-white to light yellow solid
  • Soluble in DMSO (250 mg/mL)
Catalog No. 50-004-32653 Supplier Medchemexpress LLC Supplier No. HYA0110A50MG
Cefotiam hexetil hydrochloride is an oral third-generation cephalosporin. It functions as a proagent of cefotiam, an antibiotic, though it does not possess anti-bacterial properties itself. This product is intended for research use only and is not sold to patients.
  • Functions as a proagent of cefotiam
  • Oral third-generation cephalosporin
  • Cefotiam is known as an antibiotic
  • Suitable for research use only
  • Off-white to light yellow solid
  • Soluble in DMSO (250 mg/mL)